1-(3-Bromo-2-fluoro-5-methylphenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

117372-5G 117372-1G 117372-25G 117372-500MG
77631-070EA 1484.23 USD
77631-070 77636-386 77657-230 77644-384
1-(3-Bromo-2-fluoro-5-methylphenyl)-2-fluoroethanone ≥95%
1-(3-Bromo-2-fluoro-5-methylphenyl)-2-fluoroethanone

Precaución: For Research Use Only

Formula: 249.052 g/mol
MW: C₉H₇BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD34823471
CAS Number: 2918960-84-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR