(3-Bromo-2-fluoro-5-methylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

117368-5G 117368-25G 117368-1G 117368-500MG
77631-454EA 1441 USD
77631-454 77653-270 77640-390 77644-378
(3-Bromo-2-fluoro-5-methylphenyl)trimethylsilane ≥95%
(3-Bromo-2-fluoro-5-methylphenyl)trimethylsilane

Precaución: For Research Use Only

Formula: 261.206 g/mol
MW: C₁₀H₁₄BrFSi
Storage Temperature: Ambient
MDL Number: MFCD34783455
CAS Number: 2918871-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR