Ethyl 2-(3-bromofuran-2-yl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

117296-1G 117296-5G 117296-500MG
77636-382EA 676 USD
77636-382 77657-226 77648-390
Ethyl 2-(3-bromofuran-2-yl)-2-oxoacetate ≥95%
Ethyl 2-(3-bromofuran-2-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₈H₇BrO₄
Storage Temperature: Ambient
CAS Number: 100005-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR