1-(3-Bromofuran-2-yl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

117288-500MG 117288-5G 117288-1G
77652-292EA 435 USD
77652-292 77653-268 77640-386
1-(3-Bromofuran-2-yl)-2,2,2-trifluoroethanone ≥95%
1-(3-Bromofuran-2-yl)-2,2,2-trifluoroethanone

Precaución: For Research Use Only

Formula: 242.9780 g/mol
MW: C₆H₂BrF₃O₂
Storage Temperature: Ambient
CAS Number: 1343152-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR