1-(3-Bromo-2,5-dichlorophenyl)ethanol ≥97%

Supplier: AOBChem USA

Warning

117172-5G 117172-25G 117172-1G 117172-500MG
77631-064EA 1484.23 USD
77631-064 77657-224 77640-384 77648-388
1-(3-Bromo-2,5-dichlorophenyl)ethanol ≥97%
1-(3-Bromo-2,5-dichlorophenyl)ethanol

Precaución: For Research Use Only

Formula: C₈H₇BrCl₂O
Storage Temperature: Ambient
MDL Number: MFCD32705773
CAS Number: 2921838-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR