1-(2-Bromo-4,5-difluorophenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

116935-5G 116935-25G
77633-944EA 124 USD
77633-944 77657-220
1-(2-Bromo-4,5-difluorophenyl)ethanone ≥95%
1-(2-Bromo-4,5-difluorophenyl)ethanone

Precaución: For Research Use Only

Formula: C₈H₅BrF₂O
MW: 235.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23135345
CAS Number: 127371-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR