2-Bromo-1-ethoxy-3,5-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

116584-5G 116584-1G 116584-25G 116584-500MG
77631-448EA 989 USD
77631-448 77636-378 77653-264 77644-370
2-Bromo-1-ethoxy-3,5-dimethylbenzene ≥95%
2-Bromo-1-ethoxy-3,5-dimethylbenzene

Precaución: For Research Use Only

Formula: 229.114 g/mol
MW: C₁₀H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD34783190
CAS Number: 2384713-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR