2-Bromo-1,3-diethylbenzene ≥95%

Supplier: AOBChem USA

Warning

116494-5G 116494-25G
77633-938EA 72 USD
77633-938 77653-260
2-Bromo-1,3-diethylbenzene ≥95%
2-Bromo-1,3-diethylbenzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃Br
MW: 213.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017770
CAS Number: 65232-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR