2-(2-Methoxy-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

116215-5G 116215-1G 116215-25G 116215-500MG
77631-056EA 1484.23 USD
77631-056 77636-370 77657-214 77644-366
2-(2-Methoxy-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(2-Methoxy-5-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 234.215 g/mol
MW: C₁₁H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD35165822
CAS Number: 1779804-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR