1-(2-Methoxy-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%
Supplier: AOBChem USA
|
Warning
|
116209-5G
116209-1G
116209-25G
116209-500MG
77631-052EA
1725.25
USD
77631-052
77636-368
77653-254
77648-378
1-(2-Methoxy-5-(trifluoromethyl)phenyl)cyclobutanol ≥95%
1-(2-Methoxy-5-(trifluoromethyl)phenyl)cyclobutanol
Precaución: For Research Use Only
Formula:
246.226 g/mol MW: C₁₂H₁₃F₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD34823168 CAS Number: 2921694-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...