Ethyl 2-chloro-5-methylisonicotinate ≥95%

Supplier: AOBChem USA

Warning

115768-5G 115768-1G 115768-25G 115768-500MG
77631-444EA 612 USD
77631-444 77636-364 77657-210 77648-376
Ethyl 2-chloro-5-methylisonicotinate ≥95%
Ethyl 2-chloro-5-methylisonicotinate

Precaución: For Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410741
CAS Number: 850080-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR