2-Methyl-1-(4-(trifluoromethyl)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

115480-5G 115480-1G 115480-25G 115480-500MG
77631-442EA 931 USD
77631-442 77636-358 77657-208 77644-356
2-Methyl-1-(4-(trifluoromethyl)phenyl)propan-1-one ≥95%
2-Methyl-1-(4-(trifluoromethyl)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: 216.2 g/mol
MW: C₁₁H₁₁F₃O
Storage Temperature: Ambient
MDL Number: MFCD16309307
CAS Number: 64436-58-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR