1-(2-Methoxyphenyl)-2-methylpropan-1-one ≥95%

Supplier: AOBChem USA

Warning

115463-5G 115463-1G 115463-25G 115463-500MG
77631-440EA 787 USD
77631-440 77636-356 77657-206 77644-354
1-(2-Methoxyphenyl)-2-methylpropan-1-one ≥95%
1-(2-Methoxyphenyl)-2-methylpropan-1-one

Precaución: For Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00195549
CAS Number: 74786-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR