1-(4-Bromo-1-fluoronaphthalen-2-yl)ethanone ≥95%

Supplier: AOBChem USA

Warning

115403-5G 115403-1G 115403-500MG 115403-25G
77631-044EA 1491.44 USD
77631-044 77636-354 77652-288 77653-246
1-(4-Bromo-1-fluoronaphthalen-2-yl)ethanone ≥95%
1-(4-Bromo-1-fluoronaphthalen-2-yl)ethanone

Precaución: For Research Use Only

Formula: C₁₂H₈BrFO
Storage Temperature: Ambient
MDL Number: MFCD34823091
CAS Number: 2969218-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR