1-(4-Bromo-1-fluoronaphthalen-2-yl)ethanol ≥95%

Supplier: AOBChem USA

Warning

115402-5G 115402-500MG 115402-25G 115402-1G
77631-042EA 1804.56 USD
77631-042 77652-286 77653-244 77640-372
1-(4-Bromo-1-fluoronaphthalen-2-yl)ethanol ≥95%
1-(4-Bromo-1-fluoronaphthalen-2-yl)ethanol

Precaución: For Research Use Only

Formula: 269.1096 g/mol
MW: C₁₂H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD34823090
CAS Number: 2969439-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR