Ethyl 2-(5-chloro-2-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

114772-5G 114772-1G 114772-25G 114772-500MG
77631-438EA 920 USD
77631-438 77636-352 77653-242 77648-372
Ethyl 2-(5-chloro-2-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(5-chloro-2-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 226.656 g/mol
MW: C₁₁H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD11113521
CAS Number: 1256482-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR