Ethyl 2-(3-chloro-2-methylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

114769-5G 114769-1G 114769-25G 114769-500MG
77631-436EA 661 USD
77631-436 77636-350 77657-204 77644-350
Ethyl 2-(3-chloro-2-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3-chloro-2-methylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 226.656 g/mol
MW: C₁₁H₁₁ClO₃
Storage Temperature: Ambient
MDL Number: MFCD11113531
CAS Number: 1256466-32-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR