Ethyl 2-(4-chloro-2-fluoro-3-methylphenyl)-2-oxoacetate ≥95%
Supplier: AOBChem USA
|
Warning
|
114768-5G
114768-25G
114768-500MG
114768-1G
77631-038EA
1484.23
USD
77631-038
77635-404
77652-284
77640-368
Ethyl 2-(4-chloro-2-fluoro-3-methylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(4-chloro-2-fluoro-3-methylphenyl)-2-oxoacetate
Precaución: For Research Use Only
Formula:
244.647 g/mol MW: C₁₁H₁₀ClFO₃ Storage Temperature: Ambient |
MDL Number:
MFCD26681886 CAS Number: 1510715-71-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...