Ethyl 2-oxo-2-(3-phenoxyphenyl)acetate ≥95%

Supplier: AOBChem USA

Warning

114751-5G 114751-25G 114751-1G 114751-500MG
77631-036EA 1484.23 USD
77631-036 77635-402 77640-366 77644-348
Ethyl 2-oxo-2-(3-phenoxyphenyl)acetate ≥95%
Ethyl 2-oxo-2-(3-phenoxyphenyl)acetate

Precaución: For Research Use Only

Formula: 270.28 g/mol
MW: C₁₆H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD16618264
CAS Number: 77394-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR