4-Bromo-2-chloro-6-iodophenol ≥97%

Supplier: AOBChem USA

Warning

114375-5G 114375-25G 114375-1G 114375-500MG
77631-434EA 869 USD
77631-434 77657-200 77640-362 77648-368
4-Bromo-2-chloro-6-iodophenol ≥97%
4-Bromo-2-chloro-6-iodophenol

Precaución: For Research Use Only

Formula: C₆H₃BrClIO
Storage Temperature: Ambient
MDL Number: MFCD26895843
CAS Number: 858855-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR