2-Bromo-6-fluoro-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

114368-5G 114368-1G 114368-500MG
77631-034EA 1777.78 USD
77631-034 77636-348 77648-366
2-Bromo-6-fluoro-4-iodophenol ≥97%
2-Bromo-6-fluoro-4-iodophenol

Precaución: For Research Use Only

Formula: C₆H₃BrFIO
Storage Temperature: Ambient
MDL Number: MFCD28739582
CAS Number: 1805108-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR