Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 4-[(3-fluorobenzyl)oxy]benzoate

114215-5G 114215-25G
77633-932EA 163 USD
77633-932 77653-238
Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate ≥97%
Methyl-4-[(3-fluorobenzyl)oxy]benzenecarboxylate

Precaución: For Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01568714
CAS Number: 454473-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR