2,6-Dichloro-3-iodo-5-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

113762-5G 113762-1G 113762-25G 113762-500MG
77631-032EA 1847.82 USD
77631-032 77636-346 77653-236 77644-344
2,6-Dichloro-3-iodo-5-methylpyridine ≥95%
2,6-Dichloro-3-iodo-5-methylpyridine

Precaución: For Research Use Only

Formula: 287.913 g/mol
MW: C₆H₄Cl₂IN
Storage Temperature: Ambient
MDL Number: MFCD34593600
CAS Number: 2969355-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR