2,6-Dichloro-5-methylpyridin-3-amine ≥95%

Supplier: AOBChem USA

Warning

113758-5G 113758-25G
77633-930EA 126 USD
77633-930 77653-232
2,6-Dichloro-5-methylpyridin-3-amine ≥95%
2,6-Dichloro-5-methylpyridin-3-amine

Precaución: For Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20697565
CAS Number: 58596-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR