3-Iodo-4-methylbenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

113736-5G 113736-1G 113736-25G
77633-928EA 228 USD
77633-928 77636-344 77657-198
3-Iodo-4-methylbenzotrifluoride ≥95%
3-Iodo-4-methylbenzotrifluoride

Precaución: For Research Use Only

Formula: C₈H₆F₃I
MW: 286.033 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143543
CAS Number: 13055-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR