4-Bromo-2-(difluoromethyl)-1-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

113588-5G 113588-25G 113588-1G 113588-500MG
77631-432EA 1305 USD
77631-432 77653-228 77644-342 77648-358
4-Bromo-2-(difluoromethyl)-1-iodobenzene ≥97%
4-Bromo-2-(difluoromethyl)-1-iodobenzene

Precaución: For Research Use Only

Formula: C₇H₄BrF₂I
MW: 332.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391112
CAS Number: 1261819-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR