2-Chloro-5-fluoro-3-pyridineboronic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
2-Chloro-5-fluoropyridine-3-boronic acid
11352-5G
11352-25G
11352-1G
11352-500MG
77631-430EA
684
USD
77631-430
77653-226
77640-358
77648-356
2-Chloro-5-fluoro-3-pyridineboronic acid ≥95%
2-Chloro-5-fluoro-3-pyridineboronic acid
Precaución: For Research Use Only
Formula:
C₅H₄BClFNO₂ MW: 175.35 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03094988 CAS Number: 913373-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...