5-Bromo-6-chloropicolinamide ≥97%

Supplier: AOBChem USA

Warning

113346-5G 113346-25G 113346-1G 113346-500MG
77631-428EA 712 USD
77631-428 77653-224 77640-356 77648-354
5-Bromo-6-chloropicolinamide ≥97%
5-Bromo-6-chloropicolinamide

Precaución: For Research Use Only

Formula: 235.466 g/mol
MW: C₆H₄BrClN₂O
Storage Temperature: Ambient
MDL Number: MFCD27922760
CAS Number: 1805577-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR