1,4-Dimethoxy-2-methylnaphthalene ≥97%

Supplier: AOBChem USA

Warning

113219-5G 113219-1G 113219-25G 113219-500MG
77631-028EA 1484.23 USD
77631-028 77636-340 77653-222 77644-340
1,4-Dimethoxy-2-methylnaphthalene ≥97%
1,4-Dimethoxy-2-methylnaphthalene

Precaución: For Research Use Only

Formula: C₁₃H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD12828651
CAS Number: 53772-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR