4-Bromo-2-chloro-1-((2-methylbenzyl)oxy)benzene ≥95%

Supplier: AOBChem USA

Warning

112619-5G 112619-25G 112619-1G 112619-500MG
77631-026EA 1484.23 USD
77631-026 77635-400 77640-354 77644-338
4-Bromo-2-chloro-1-((2-methylbenzyl)oxy)benzene ≥95%
4-Bromo-2-chloro-1-((2-methylbenzyl)oxy)benzene

Precaución: For Research Use Only

Formula: 311.601 g/mol
MW: C₁₄H₁₂BrClO
Storage Temperature: Ambient
MDL Number: MFCD34781628
CAS Number: 2918924-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR