1-Bromo-3-chloro-4-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

112413-1G 112413-500MG
77633-926EA 1596 USD
77633-926 77644-336
1-Bromo-3-chloro-4-fluoro-2-methylbenzene ≥97%
1-Bromo-3-chloro-4-fluoro-2-methylbenzene

Precaución: For Research Use Only

Formula: 223.47 g/mol
MW: C₇H₅BrClF
Storage Temperature: Ambient
MDL Number: MFCD28734051
CAS Number: 1785149-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR