2,6-Bis(benzyloxy)nicotinic acid ≥97%

Supplier: AOBChem USA

Warning

112389-5G 112389-1G 112389-500MG
77631-024EA 1484.23 USD
77631-024 77636-338 77644-334
2,6-Bis(benzyloxy)nicotinic acid ≥97%
2,6-Bis(benzyloxy)nicotinic acid

Precaución: For Research Use Only

Formula: 335.353 g/mol
MW: C₂₀H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD34781530
CAS Number: 16727-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR