4-Bromo-2,3,5-trifluoropyridine ≥95%

Supplier: AOBChem USA

Warning

11203-5G 11203-1G 11203-25G 11203-500MG
77631-426EA 889 USD
77631-426 77636-336 77657-196 77648-352
4-Bromo-2,3,5-trifluoropyridine ≥95%
4-Bromo-2,3,5-trifluoropyridine

Precaución: For Research Use Only

Formula: 211.967 g/mol
MW: C₅HBrF₃N
Storage Temperature: Ambient
MDL Number: MFCD08436092
CAS Number: 851178-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR