2-(2,4-Dibromophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

111799-5G 111799-1G 111799-25G 111799-500MG
77631-022EA 1484.23 USD
77631-022 77636-334 77653-220 77648-350
2-(2,4-Dibromophenyl)-1,3-dioxolane ≥97%
2-(2,4-Dibromophenyl)-1,3-dioxolane

Precaución: For Research Use Only

Formula: 307.967 g/mol
MW: C₉H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22050781
CAS Number: 331714-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR