(3-(Cyclohexylmethoxy)-5-fluorophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

111605-500MG 111605-5G 111605-1G
77652-282EA 398 USD
77652-282 77657-194 77640-352
(3-(Cyclohexylmethoxy)-5-fluorophenyl)boronic acid ≥97%
(3-(Cyclohexylmethoxy)-5-fluorophenyl)boronic acid

Precaución: For Research Use Only

Formula: C₁₃H₁₈BFO₃
MW: 252.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23089552
CAS Number: 2246679-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR