4-Bromo-2,3-difluoro-(n-hexyloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

11160-5G 11160-1G 11160-25G 11160-500MG
77631-424EA 793 USD
77631-424 77636-332 77653-218 77648-346
4-Bromo-2,3-difluoro-(n-hexyloxy)benzene ≥97%
4-Bromo-2,3-difluoro-(n-hexyloxy)benzene

Precaución: For Research Use Only

Formula: 293.148 g/mol
MW: C₁₂H₁₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD29043895
CAS Number: 256383-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR