4-Butoxy-2-chlorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

111536-5G 111536-25G 111536-1G 111536-500MG
77631-422EA 1055 USD
77631-422 77653-216 77640-350 77644-332
4-Butoxy-2-chlorobenzaldehyde ≥97%
4-Butoxy-2-chlorobenzaldehyde

Precaución: For Research Use Only

Formula: C₁₁H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD34781055
CAS Number: 1692304-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR