4-Bromo-3-(trifluoromethoxy)benzal fluoride ≥95%

Supplier: AOBChem USA

Warning

111282-5G 111282-25G 111282-1G 111282-500MG
77633-924EA 507 USD
77633-924 77657-192 77640-348 77648-344
4-Bromo-3-(trifluoromethoxy)benzal fluoride ≥95%
4-Bromo-3-(trifluoromethoxy)benzal fluoride

Precaución: For Research Use Only

Formula: C₈H₄BrF₅O
MW: 291.013 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812119
CAS Number: 2167351-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR