4-Bromo-3-chloro-2-methylbenzonitrile ≥95%

Supplier: AOBChem USA

Warning

111232-1G 111232-5G 111232-500MG
77636-330EA 530 USD
77636-330 77657-190 77644-330
4-Bromo-3-chloro-2-methylbenzonitrile ≥95%
4-Bromo-3-chloro-2-methylbenzonitrile

Precaución: For Research Use Only

Formula: C₈H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD19686177
CAS Number: 1349716-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR