2-Fluoro-6-(methylthio)nicotinic acid ≥95%

Supplier: AOBChem USA

Warning

111194-500MG 111194-5G 111194-1G
77652-280EA 477 USD
77652-280 77653-212 77640-344
2-Fluoro-6-(methylthio)nicotinic acid ≥95%
2-Fluoro-6-(methylthio)nicotinic acid

Precaución: For Research Use Only

Formula: C₇H₆FNO₂S
Storage Temperature: Ambient
MDL Number: MFCD34780961
CAS Number: 2918836-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR