3-Bromo-5-chloro-4-iodophenol ≥95%

Supplier: AOBChem USA

Warning

111045-1G 111045-500MG 111045-5G
77636-328EA 528 USD
77636-328 77652-278 77653-210
3-Bromo-5-chloro-4-iodophenol ≥95%
3-Bromo-5-chloro-4-iodophenol

Precaución: For Research Use Only

Formula: 333.349 g/mol
MW: C₆H₃BrClIO
Storage Temperature: Ambient
MDL Number: MFCD34780918
CAS Number: 2091672-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR