2-(Benzyloxy)-5-bromo-3-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

110842-5G 110842-25G
77633-922EA 154 USD
77633-922 77653-208
2-(Benzyloxy)-5-bromo-3-methylpyridine ≥95%
2-(Benzyloxy)-5-bromo-3-methylpyridine

Precaución: For Research Use Only

Formula: C₁₃H₁₂BrNO
Storage Temperature: Ambient
MDL Number: MFCD21604769
CAS Number: 1289270-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR