2-Isopropoxy-6-(methylthio)pyridine ≥95%

Supplier: AOBChem USA

Warning

110758-5G 110758-25G 110758-1G 110758-500MG
77631-020EA 1484.23 USD
77631-020 77657-188 77640-342 77648-342
2-Isopropoxy-6-(methylthio)pyridine ≥95%
2-Isopropoxy-6-(methylthio)pyridine

Precaución: For Research Use Only

Formula: 183.271 g/mol
MW: C₉H₁₃NOS
Storage Temperature: Ambient
MDL Number: MFCD34780786
CAS Number: 2918849-09-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR