5-Bromo-4-methyl-2-(propylthio)pyridine ≥97%

Supplier: AOBChem USA

Warning

110675-5G 110675-25G 110675-1G 110675-500MG
77631-418EA 1034 USD
77631-418 77657-186 77640-340 77644-326
5-Bromo-4-methyl-2-(propylthio)pyridine ≥97%
5-Bromo-4-methyl-2-(propylthio)pyridine

Precaución: For Research Use Only

Formula: C₉H₁₂BrNS
Storage Temperature: Ambient
MDL Number: MFCD34780748
CAS Number: 1602959-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR