2,3,5-Trichloro-6-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

110106-5G 110106-25G 110106-1G 110106-500MG
77633-918EA 291 USD
77633-918 77653-202 77640-338 77648-338
2,3,5-Trichloro-6-methoxypyridine ≥97%
2,3,5-Trichloro-6-methoxypyridine

Precaución: For Research Use Only

Formula: C₆H₄Cl₃NO
MW: 212.46 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01940632
CAS Number: 31557-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR