2-(2,6-Difluoro-3-iodophenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

110079-5G 110079-1G 110079-25G 110079-500MG
77631-018EA 1484.23 USD
77631-018 77636-324 77653-200 77648-336
2-(2,6-Difluoro-3-iodophenyl)propan-2-ol ≥95%
2-(2,6-Difluoro-3-iodophenyl)propan-2-ol

Precaución: For Research Use Only

Formula: C₉H₉F₂IO
Storage Temperature: Ambient
CAS Number: 3031788-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR