4-(5-Chloro-2-methoxypyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥95%
Supplier: AOBChem USA
|
Warning
|
109388-5G
109388-25G
109388-1G
109388-500MG
77631-016EA
1484.23
USD
77631-016
77635-398
77640-334
77648-332
4-(5-Chloro-2-methoxypyridin-4-yl)tetrahydro-2H-pyran-4-ol ≥95%
4-(5-Chloro-2-methoxypyridin-4-yl)tetrahydro-2H-pyran-4-ol
Precaución: For Research Use Only
Formula:
C₁₁H₁₄ClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD34780147 CAS Number: 2918912-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...