5-Bromo-6-chloropicolinonitrile ≥95%

Supplier: AOBChem USA

Warning

109308-5G 109308-1G 109308-25G 109308-500MG
77633-914EA 287 USD
77633-914 77636-322 77653-198 77644-324
5-Bromo-6-chloropicolinonitrile ≥95%
5-Bromo-6-chloropicolinonitrile

Precaución: For Research Use Only

Formula: C₆H₂BrClN₂
MW: 217.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18257828
CAS Number: 1256823-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR