2-Bromo-5-chloro-1,3-diisopropylbenzene ≥95%

Supplier: AOBChem USA

Warning

108986-5G 108986-1G 108986-500MG
77631-014EA 1484.23 USD
77631-014 77640-332 77644-322
2-Bromo-5-chloro-1,3-diisopropylbenzene ≥95%
2-Bromo-5-chloro-1,3-diisopropylbenzene

Precaución: For Research Use Only

Formula: 275.612 g/mol
MW: C₁₂H₁₆BrCl
Storage Temperature: Ambient
MDL Number: MFCD31693031
CAS Number: 2143028-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR