Tert-butyl 3-fluoro-2,4-dimethylbenzoate ≥95%

Supplier: AOBChem USA

Warning

108355-5G 108355-25G 108355-1G 108355-500MG
77631-012EA 1740.7 USD
77631-012 77635-396 77636-318 77648-328
Tert-butyl 3-fluoro-2,4-dimethylbenzoate ≥95%
Tert-butyl 3-fluoro-2,4-dimethylbenzoate

Precaución: For Research Use Only

Formula: 224.271 g/mol
MW: C₁₃H₁₇FO₂
Storage Temperature: Ambient
MDL Number: MFCD34779806
CAS Number: 2918839-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR